Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55265

Millipore

Listeria Motility Medium

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Motility Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

Agency

according to ISO 11290-1:2017
according to ISO 11290-2:2017

Quality Level

sterility

non-sterile

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 3.5 g/L
casein enzymic hydrolysate, 20.0 g/L
peptic digest of animal tissue, 6.1 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | growth & motility: suitable
microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages

microbiology

suitability

nonselective for Listeria spp.

Application

Listeria Motility Medium is a semisolid medium recommended by ISO Committee for testing motility of Listeria monocytogenes.

Preparation Note

Suspend 29.6 grams in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Dispense in tubes and sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Allow the medium to cool in an upright position.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Sun Lee et al.
Virology, 529, 43-56 (2019-01-21)
The large GTPase dynamin 2 controls both endosomal fission and microtubule acetylation. Here we report that dynamin 2 alters microtubules and regulates the trafficking of human adenovirus type 37. Dynamin 2 knockdown by siRNA in infected cells resulted in accumulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service