Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABN1838

Sigma-Aldrich

Anti-AGE Antibody, Methylglyoxal Antibody, clone 3D11

clone 3D11, from mouse

Synonym(s):

Methylglyoxal, AGE Product methylglyoxal, clone 3D11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D11, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human, rat

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG2aλ

shipped in

ambient

target post-translational modification

unmodified

Related Categories

General description

Chronic accumulation of Advanced Glycation End products (AGEs) is a common feature during aging, hyperlipidemia, and diabetes and affects function of proteins, lipids, and DNA. Although formation of AGE products is considered to be predominantly an endogenous process, certain external factors, such as some dietary products and smoking can increase the level of AGE in tissues. Under certain conditions glucose may form covalent adducts with the plasma proteins through the process of glycation. Glycation is a non-enzymatic reaction of reducing carbohydrates with lysine side chains and N-terminal amino groups of proteins. The term AGE often refers to non-reactive terminal products, such as N- carboxymethyllysine (CML) as well as reactive precursors, such as methylglyoxal (MG), which is generated through a non-oxidative process from triose phosphates during anaerobic glycolysis. MG reacts with arginine, lysine, and cysteine residues in proteins to form AGE products. Excessive glycation can disrupt protein function by altering enzyme activity, receptor function, and molecular conformation. Protein glycation and subsequent formation of AGE products contributes to the pathogenesis of diabetes-related complications, such as retinopathy, nephropathy, and neuropathy. AGE products interact with a variety of cell-surface AGE-binding receptors (RAGE) that results in their endocytosis and degradation. It can also generate pro-oxidant and pro-inflammatory events in cells. AGE products could also contribute to the development or worsening of many degenerative diseases, including Alzheimer′s disease. (Ref.: Cai,W., et al. (2002). Mol. Med 8 (7): 337-346).

Specificity

Clone 3D11 specifically reacts with methylglyoxal modified proteins. It does not react with CML, CEL, or other AGE epitopes.

Immunogen

Methylglyoxal-cojugated Ovalbumin.

Application

Anti-AGE, Methylglyoxal, clone 3D11, Cat. No. MABN1838, is a highly specific mouse monoclonal antibody that targets Methylglyoxal modified proteins and has been tested in ELISA and Immunohistochemistry (Paraffin).
ELISA Analysis: A representative lot detected AGE, Methylglyoxal in ELISA applications (Vlassara, H., et. al. (2002). Proc Natl Acad Sci USA. 99(24):15596-601; Cai, W., et. al. (2008). Am J Pathol. 173(2):327-36; Cai, W., et. al. (2012). Proc Natl Acad Sci USA. 109(39):15888-15893).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry in mouse kidney tissue.

Immunohistochemistry Analysis: A 1:3,000 dilution of this antibody detected AGE, Methylglyoxal in kidney tissue from mice fed high AGE diets.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service