Skip to Content
MilliporeSigma

MAK467

Aspartate Transaminase (AST) Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

AST Activity Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12352204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

application(s)

pharmaceutical

detection method

colorimetric

relevant disease(s)

hematological disorder; cardiovascular diseases; musculoskeletal disorders; genitourinary disorders; gastrointestinal diseases

shipped in

wet ice

storage temp.

−20°C

General description

Aspartate Transaminase (AST), also known as serum glutamic oxaloacetic transaminase (GOT) or aspartate aminotransferase (ASAT/AAT), facilitates the conversion of aspartate and α-ketoglutarate to oxaloacetate and glutamate. There are two isoenzymes in humans: GOT1 is a cytosolic isoenzyme derived from red blood cells and heart; GOT2 is the mitochondrial isoenzyme found mainly in the liver. AST is elevated in liver and muscle diseases. It is part of diagnostic testing for liver function, myocardial infarction, acute pancreatitis, acute hemolytic anemia, severe burns, acute renal disease, and trauma.

Application

The kit is suitable for AST activity determination in serum, plasma, and other biological samples, as well as for studying the effects of drugs on AST activity.

Biochem/physiol Actions

The Aspartate Transaminase (AST) Assay Kit is based on the quantification of oxaloacetate produced by AST. In this assay, oxaloacetate and NADH are converted to malate and NAD+ by the enzyme malate dehydrogenase. The decrease in NADH absorbance at 340 nm is proportionate to AST activity.

Features and Benefits

  • Linear detection range: 2 to 100 U/L
  • This simple, convenient assay can be carried out in a microplate or a cuvette and takes only 10 minutes



Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library