Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN1813

Sigma-Aldrich

Anti-sodium/hydrogen exchanger 3 Antibody, clone 3H3

clone 3H3, from mouse

Synonym(s):

Sodium/hydrogen exchanger 3, Na(+)/H(+) exchanger 3, Solute carrier family 9 member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H3, monoclonal

species reactivity

human, mouse, rat, opossum

species reactivity (predicted by homology)

porcine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Slc9A7(236727)

General description

Sodium/hydrogen exchanger 3 (UniProt: Q28362; also known as Na(+)/H(+) exchanger 3, NHE-3, Solute carrier family 9 member 3) is encoded by the SLC9A3 (also known as NHE3) gene in Didelphis virginiana (North American opossum). NHE-3 is a multi-pass membrane protein that is involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. NHE-3 is predominantly expressed in kidney and gut, where it is found on the apical membranes of polarized epithelial cells and plays a central role in transepithelial reabsorption of sodium ions and secretion of hydrogen ions. Renal NHE-3 is essential in maintaining blood pressure and steady-state plasma sodium levels when dietary sodium chloride intake is modified. In the intestine epithelial cells, it localizes to the ileal brush border. Dysfunction of NHE-3 in intestine is associated with a variety of diarrheal diseases. HNE-3 can be phosphorylated by SGK1 at Ser669 that increases its abundance at the cell membrane. (Ref.: D Souza, S et al. (1998). J. Biol. Chem. 273(4); 2035 2043; Fenton, RA et al (2017). Kidney Int. doi: 10.1016/j.kint.2017.02.001. [Epub ahead of print]).

Specificity

Clone CH3 detects Sodium/hydrogen exchanger 3 in human, opossum, and rat kidney cells. It targets an epitope within 356 amino acids from the C-terminal half.

Immunogen

MBP-conjugated recombinant fragment corresponding to 356 amino acids from the C-terminal half of Opossum sodium/hydrogen exchanger 3.

Application

Anti-NHE3, clone 3H3, Cat. No. MABN1813, is a highly specific mouse monoclonal antibody that targets Sodium/hydrogen exchanger 3 and has been tested for use in Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected NHE3 in human kidney and human stomach tissue sections.

Western Blotting Analysis: A representative lot detected NHE3 in Western Blotting applications (Wang, D., et. al. (2006). Am J Physiol Renal Physiol. 290(2):F428-37; Queiroz-Leite, G.D., et. al. (2011). Biochem Biophys Res Commun. 409(3):470-6; Goyal, S., et. al. (2005). Am J Physiol Renal Physiol. 288(3):F530-8; Kocinsky, H.S., et. al. (2005). Am J Physiol Renal Physiol. 289(2):F249-58; Di Sole, F., et. al. (2008). J Cell Physiol. 216(1):221-33; Baum, M., et. al. (2012). Am J Physiol Renal Physiol. 303(11):F1495-502; Lessa, L.M., et. al. (2012). Am J Physiol Renal Physiol. 303(10):F1399-408).

Immunohistochemistry Analysis: A representative lot detected NHE3 in Immunohistochemistry applications (Pao, A.C., et. al. (2010). Am J Physiol Renal Physiol. 299(6):F1496-506).

Immunoprecipitation Analysis: A representative lot detected NHE3 in Immunoprecipitation applications (Girardi, A.C., et. al. (2004). Am J Physiol Renal Physiol. 287(5):C1238-45).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in COS-7 cells transfected with wild type NHE3 plasmid DNA.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected NHE3 in lysates from COS-7 cells transfected with wild type NHE3 plasmid DNA.

Target description

~75 kDa observed; 94.77 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan J Adam et al.
Kidney360, 1(10), 1105-1115 (2021-07-16)
The 5/6 nephrectomy (5/6Nx) rat model recapitulates many elements of human CKD. Within weeks of surgery, 5/6Nx rats spontaneously exhibit proximal tubular damage, including the production of very large extracellular vesicles and brush border shedding. We hypothesized that production and
Yanli Dong et al.
Science advances, 8(21), eabn3925-eabn3925 (2022-05-26)
Sodium-proton exchanger 3 (NHE3/SLC9A3) located in the apical membrane of renal and gastrointestinal epithelia mediates salt and fluid absorption and regulates pH homeostasis. As an auxiliary regulatory factor of NHE proteins, calcineurin B homologous protein 1 (CHP1) facilitates NHE3 maturation
Md Kaimul Ahsan et al.
Journal of clinical medicine, 11(14) (2022-07-28)
Microvillus inclusion disease (MVID), a lethal congenital diarrheal disease, results from loss of function mutations in the apical actin motor myosin VB (MYO5B). How loss of MYO5B leads to both malabsorption and fluid secretion is not well understood. Serum glucocorticoid-inducible
Md Kaimul Ahsan et al.
American journal of physiology. Gastrointestinal and liver physiology, 319(2), G121-G132 (2020-06-23)
Nongenomic glucocorticoid (GC) and serum- and glucocorticoid-inducible kinase 1 (SGK1) signaling regulate ion transport, but CFTR has not been investigated in the intestine. We examined GC, SGK1, and phosphatidylinositol 3-kinase (PI3K) kinase signaling of CFTR ion transport in native intestine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service