Skip to Content
MilliporeSigma

04-148

Anti-CLEC9A Antibody, clone 10B4

clone 10B4, from rat

Synonym(s):

C-type lectin domain family 9 member A, Dendritic cell natural killer lectin group receptor 1, CLEC9A, CLEC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
10B4, monoclonal
Application:
FACS, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10B4, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Clec9A(232414)

General description

CLEC9A, also known as Dendritic cell natural killer lectin group receptor 1 (DNGR-1), and encoded by the gene Clec9a/Dngr-1, functions as an endocytic receptor on a small subset of myeloid cells specialized for the uptake and processing of material from dead cells. CLEC9A functions as a SYK-coupled C-type lectin receptor to mediate sensing of necrosis by the principal dendritic-cell subset involved in regulating cross-priming to cell-associated antigens. CLEC9A recognizes filamentous form of actin in association with particular actin-binding domains of cytoskeletal proteins, including spectrin, exposed when cell membranes are damaged, and mediates the cross-presentation of dead-cell associated antigens in a Syk-dependent manner. CLEC9A is localized to the plasma membrane. CLEC9A is expressed in dendritic cells and has multiple isoforms; isoform 4 is expressed at high levels by CD8+ dendritic cells and at low levels by plasmacytoid DCs but not by other hematopoietic cells.
~27 kDa and ~16 kDa observed. Uniprot describes five isoforms at 27 kDa, 27 kDa, 30 kDa, 20 kDa, and 17 kDa

Immunogen

KLH-conjugated linear peptide corresponding to mouse CLEC9A.

Application

Detect CLEC9A using this rat monoclonal antibody, Anti-CLEC9A Antibody, clone 10B4 validated for use in western blotting & Flow Cytometry.
Flow Cytometry Analysis: A representative lot from an independent laboratory detected CLEC9A in mouse Conventional Dendritic cells and on most Plasmacytoid Dendritic cells (Caminschi, I., et al. (2008). Blood. 112(8):3264-3273.).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in mouse lymph node tissue lysate.

Western Blotting Analysis: 1 μg/mL of this antibody detected CLEC9A in 10 μg of mouse lymph node tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library