Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAB3136

Sigma-Aldrich

Anti-Na+/H+ Exchanger-3 Antibody, isoform NHE3, clone 4F5

clone 4F5, Chemicon®, from mouse

Synonym(s):

NHE3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F5, monoclonal

species reactivity

rat, rabbit

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Na+/H+ exchanger, isoform NHE3.

Immunogen

Epitope: isoform NHE3
Maltose Binding Protein fusion protein containing C-terminal 131 amino acids of rabbit NHE3.

Application

Anti-Na+/H+ Exchanger-3 Antibody, isoform NHE3, clone 4F5 detects level of Na+/H+ Exchanger-3 & has been published & validated for use in IC, IH(P), IP & WB.
Immunoblotting (rabbit 80 kDa and rat ~83 kDa)

Immunohistochemistry (rat) on PLP or paraformaldehyde fixed tissue. Works on paraffin embedded tissue sections.

Immunocytochemistry

Immunoprecipitation (use non-ionic detergent)

Optimal working dilutions must be determined by the user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Linkage

Replaces: MAB3134

Physical form

Format: Purified
Liquid in 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanling Yan et al.
Molecular and cellular biochemistry, 367(1-2), 175-183 (2012-05-24)
We have demonstrated that ouabain regulates protein trafficking of the Na/K-ATPase α1 subunit and NHE3 (Na/H exchanger, isoform 3) via ouabain-activated Na/K-ATPase signaling in porcine LLC-PK1 cells. To investigate whether this mechanism is species-specific, ouabain-induced regulation of the α1 subunit
Elevated BSC-1 and ROMK expression in Dahl salt-sensitive rat kidneys.
Hoagland, KM; Flasch, AK; Dahly-Vernon, AJ; dos Santos, EA; Knepper, MA; Roman, RJ
Hypertension null
Novel signaling mechanisms of intracellular angiotensin II-induced NHE3 expression and activation in mouse proximal tubule cells.
Li, XC; Hopfer, U; Zhuo, JL
American Journal of Physiology: Renal Physiology null
Xiaochun Li et al.
Cells, 12(11) (2023-06-10)
The current prevailing paradigm in the renin-angiotensin system dictates that most, if not all, biological, physiological, and pathological responses to its most potent peptide, angiotensin II (Ang II), are mediated by extracellular Ang II activating its cell surface receptors. Whether
Brandon A Kemp et al.
Journal of the American Heart Association, 8(9), e012016-e012016 (2019-05-02)
Background Previous studies demonstrated that angiotensin (Ang) III , not Ang II , is the predominant endogenous agonist for Ang type-2 receptor ( AT 2R)-induced natriuresis in normal rats, and that hypertensive 12-week-old spontaneously hypertensive rats ( SHR ) lack

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service