Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN102

Sigma-Aldrich

Anti-ARX Antibody, clone 11F6.2

clone 11F6.2, from mouse

Synonym(s):

Homeobox protein ARX, Aristaless-related homeobox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11F6.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ARX(170302)

General description

Aristaless-related homeobox protein (ARX) is a transcription factor that plays an important role in the development of the brain, testis, and pancreas. The gene targets and nuclear associations of ARX are less clear, although it has been demonstrated that ARX interacts with the Pax4 promoter. However, over fifty ARX mutations have been identified and the resultant phenotypes have been well studied. ARX mutations are X-linked and lead to various syndromes including X-linked West syndrome; X-linked myoclonic epilepsy; Partington syndrome; and X-linked mental retardation.

Immunogen

GST-tagged recombinant protein corresponding to human ARX.

Application

Detect ARX using this Anti-ARX Antibody, clone 11F6.2 validated for use in Western Blotting, IHC(P).
Immunohistochemistry Analysis: A 1:1,000 dilution from a reprsentative lot detected ARX in human brain tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in human pancreas tissue lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected ARX in 200 µg of human pancreas tissue lysate.

Target description

~58 kDa observed. An uncharacterized band may be observed at ~23 kDa in some tissue lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human pancreas tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenzel M Hackeng et al.
Diagnostic cytopathology, 48(4), 308-315 (2019-12-18)
The transcription factors ARX and PDX1, and alternative lengthening of telomeres (ALT) were recently described as prognostic markers for resected non-functional pancreatic neuroendocrine tumors (PanNETs). ALT positive tumors with ARX expression relapse most often. Currently, tumor size is the only
Wenzel M Hackeng et al.
Endocrine pathology, 31(2), 108-118 (2020-02-28)
Insulin-producing pancreatic neuroendocrine tumors (PanNETs)/insulinomas are generally considered to be indolent tumors with an excellent prognosis after complete resection. However, some insulinomas have a poor prognosis due to relapses and metastatic disease. Recently, studies in non-functional PanNETs indicated that behavior
Wenzel M Hackeng et al.
Gut (2021-04-15)
Recent studies have found aristaless-related homeobox gene (ARX)/pancreatic and duodenal homeobox 1 (PDX1), alpha-thalassemia/mental retardation X-linked (ATRX)/death domain-associated protein (DAXX) and alternative lengthening of telomeres (ALT) to be promising prognostic biomarkers for non-functional pancreatic neuroendocrine tumours (NF-PanNETs). However, they have
Francesca Cinti et al.
The Journal of clinical endocrinology and metabolism, 101(3), 1044-1054 (2015-12-30)
Diabetes is associated with a deficit of insulin-producing β-cells. Animal studies show that β-cells become dedifferentiated in diabetes, reverting to a progenitor-like stage, and partly converting to other endocrine cell types. To determine whether similar processes occur in human type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service