Skip to Content
MilliporeSigma

1.07267

SAFC

L-Phenylalanine

EMPROVE® EXPERT, Ph. Eur., JP, USP

Pharma Manufacturing

Synonym(s):

L-Phenylalanine, Phe, 2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
165.19
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-568-1

Agency

JP
Ph. Eur.
USP

Quality Level

vapor pressure

<1 Pa ( 25 °C)

description

EMPROVE® EXPERT Ph Eur,JP,USP

product line

EMPROVE® EXPERT

form

solid

pH

5.4-6.0 (20 °C, 10 g/L in H2O)

mp

275-283 °C (decomposition)

solubility

27 g/L

density

1.34 g/cm3 at 20 °C

bulk density

580 kg/m3

application(s)

parenterals

storage temp.

2-30°C

SMILES string

[N+H3]C(Cc1ccccc1)C(=O)[O-]

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)

InChI key

COLNVLDHVKWLRT-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

This overview describes how use of viscosity-reducing excipients can address viscosity challenges in high concentration tangential flow filtration (TFF) and improve process economics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service