Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46-713MAG

Millipore

MILLIPLEX® MAP ß-Tubulin Total Magnetic Bead MAPmate - Cell Signaling Single Plex or Multiplex Assay

Synonym(s):

Cell Signaling MAPmate, Magnetic Bead MAPmate, ß-Tubulin Magnetic Bead Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

species reactivity

mouse, rat, human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

NCBI accession no.

UniProt accession no.

compatibility

configured for Single Plex

detection method

fluorometric (Luminex xMAP)

General description

The MILLIPLEX MAP Total ß-Tubulin Magnetic Bead MAPmates pair is used in conjunction with the MILLIPLEX MAP Cell Signaling Buffer and Detection Kit to detect the presence of total b-Tubulin in cell lysates using the Luminex® 100 IS, 200, or HTS system. Each MAPmates pair is ordered individually and may be combined for simultaneous multiplex analysis of cellular events. The MILLIPLEX MAP Cell Signaling Buffer and Detection Kit is ordered separately and consists of a common set of reagents needed for performing MAPmates assays. The detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures. Each kit contains sufficient reagents for 100 individual assays. The MILLIPLEX MAP HeLa Cell Lysate: Unstimulated included in the MILLIPLEX MAP Cell Signaling Buffer and Detection Kit may be utilized as a unstimulated control for this target.

Application

MAPmate single plexes allow you to design your own cell signaling panel. The analyte available for this kit is β-Tubulin.
Research Category
Cell Signaling
Used to detect/quantify:β-Tubulin

Components

MILLIPLEX MAP Anti- total β-Tubulin, Biotin (20X)
MILLIPLEX MAP Anti-β-Tubulin Magnetic Beads (20X)

Storage and Stability

Recommended storage for kit components is 2 - 8°C

Legal Information

Luminex is a registered trademark of Luminex Corp
MAPMATE is a trademark of Merck KGaA, Darmstadt, Germany
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Exploring genotoxicity and DNA damage through multiplexing with MILLIPLEX® multiplex genotoxicity assays using Luminex® xMAP® technology enables the high-throughput measurement of phosphorylation levels of multiple proteins simultaneously and reduces sample volume, time, and cost.

Exploring genotoxicity and DNA damage through multiplexing with MILLIPLEX® multiplex genotoxicity assays using Luminex® xMAP® technology enables the high-throughput measurement of phosphorylation levels of multiple proteins simultaneously and reduces sample volume, time, and cost.

Exploring genotoxicity and DNA damage through multiplexing with MILLIPLEX® multiplex genotoxicity assays using Luminex® xMAP® technology enables the high-throughput measurement of phosphorylation levels of multiple proteins simultaneously and reduces sample volume, time, and cost.

Exploring genotoxicity and DNA damage through multiplexing with MILLIPLEX® multiplex genotoxicity assays using Luminex® xMAP® technology enables the high-throughput measurement of phosphorylation levels of multiple proteins simultaneously and reduces sample volume, time, and cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service