Skip to Content
MilliporeSigma

AB2301

Anti-Calnexin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Calnexin, IP90, Major histocompatibility complex class I antigen-binding protein p88, p90

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, WB
Citations:
31
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human, horse, pig

species reactivity (predicted by homology)

equine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CANX(821)

General description

Calnexin is a Ca++ binding protein found in ER transmembranes that binds to specific n-linked oligosaccharides found on newly synthesized proteins. Calnexin associates with the ER membrane phosphoglycoprotein, signal sequence receptor SSRμ, and has been identified as the ER protein associated with major histocompatibility complex (MHC) class I heavy chain and with the T-cell receptor and membrane immunoglobulin before assembly of these multimeric complexes. Hence, calnexin may represent a molecular chaperone in the ER membrane that is selective for incompletely folded oligomeric membrane proteins newly translocated to the ER.
~70 kDa observed

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human Calnexin.

Application

Anti-Calnexin Antibody is an antibody against Calnexin for use in WB, IC, IH(P).
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Calnexin in NIH/3T3 cells.

Immunohistochemistry Analysis: A 1:1,000-2,000 dilution from a representative lot detected Calnexin in human brain and human prostate tissues.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Biochem/physiol Actions

This antibody recognizes Calnexin at the C-terminus.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human brain tissue lysate
Evaluated by Western Blot in human brain tissue lysate.

Western Blot Analysis: A 1:10,000 dilution of this antibody detected Calnexin on 10 µg of human brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library