Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3501123

Sigma-Aldrich

Anti-SARS-CoV-2 (COVID-19) Nucleocapsid antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

SARS coronavirus

concentration

1 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

SARS coronavirus ... N(43740575)

Looking for similar products? Visit Product Comparison Guide

General description

Coronavirus disease 2019 (COVID-19), formerly known as 2019-nCoV acute respiratory disease, is an infectious disease caused by SARS-CoV-2, a virus closely related to the SARS virus. The disease is the cause of the 2019–20 coronavirus outbreak. SARS-CoV-2 is the seventh member of the enveloped, positive-stranded RNA viruses that are able to infect humans. The SARS-CoV-2 genome, like other coronaviruses, encodes for multiple structural and nonstructural proteins. The structural proteins include spike protein (S), envelope protein (E), membrane glycoprotein (M), nucleocapsid phosphoprotein (N), and the nonstructural proteins include open reading frame 1ab (ORF1ab), ORF3a, ORF6, ORF7a, ORF8, and ORF10. Nucleocapsid (N) protein is the most abundant protein of coronavirus. It is also one of the major structural proteins and is involved in the transcription and replication of viral RNA, packaging of the encapsidated genome into virions , and interference with cell cycle processes of host cells . Moreover, in many coronaviruses, including SARS-CoV, the N protein has high immunogenic activity and is abundantly expressed during infection. It can be detected in various patient samples including nasopharyngeal aspirate, urine, and fecal. Both S and N proteins may be potential antigens for serodiagnosis of COVID-19, just as many diagnostic methods have been developed for diagnosing SARS based on S and/or N proteins.

Immunogen

Anti-SARS-CoV-2 (COVID-19) Nucleocapsid antibody was raised against a peptide corresponding to 17 amino acids near the center of SARS-CoV-2 (COVID-19) Nucleocapsid protein. The immunogen is located within 230-280 amino acids of SARS-CoV-2 (COVID-19) Nucleocapsid protein.

Application

SARS-CoV-2 (COVID-19) Nucleocapsid antibody can be used for the detection of SARS-CoV-2 (COVID-19) Nucleocapsid protein in ELISA. It will detect 4 ng of free peptide at 1 μg/mL. All other applications and species not yet tested.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service