MilliporeSigma
  • Uterine WNTS modulates fibronectin binding activity required for blastocyst attachment through the WNT/CA2+ signaling pathway in mice.
Uterine WNTS modulates fibronectin binding activity required for blastocyst attachment through the WNT/CA2+ signaling pathway in mice.
Reproductive biology and endocrinology : RB&E (2023-09-16)
Yuefei Lou, Laurie Pinel, Daniel Dufort
ABSTRACT
Adhesion of the implanting blastocyst involves the interaction between integrin proteins expressed by trophoblast cells and components present in the basement membrane of the endometrial luminal epithelium. Although several factors regulating integrins and their adhesion to fibronectin are already known, we showed that Wnt signaling is involved in the regulation of blastocyst adhesion through the trafficking of integrins expressed by trophoblast cells. Localization of Itgα5β1 by immunofluorescence and FN-binding assays were conducted on peri-implantation blastocysts treated with either Wnt5a or Wnt7a proteins. Both Wnt5a and Wnt7a induced a translocation of Itgα5β1 at the surface of the blastocyst and an increase in FN-binding activity. We further demonstrated that uterine fluid is capable of inducing integrin translocation and this activity can be specifically inhibited by the Wnt inhibitor sFRP2. To identify the Wnt signaling pathway involved in this activity, blastocysts were incubated with inhibitors of either p38MAPK, PI3K pathway or CamKII prior to the addition of Wnts. Whereas inhibition of p38MAPK and PI3K had not effect, inhibition of CamKII reduced FN-binding activity induced by Wnts. Finally, we demonstrated that inhibition of Wnts by sFRP2 reduced the binding efficiency of the blastocyst to uterine epithelial cells. Our findings provide new insight into the mechanism that regulates integrin trafficking and FN-binding activity and identifies Wnts as a key player in blastocyst attachment to the uterine epithelium.
Sigma-Aldrich
Anti-Integrin α5β1 Antibody, clone BMB5, ascites fluid, clone BMB5, Chemicon®