Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB2148-C

Sigma-Aldrich

Anti-PECAM-1 Antibody, clone P2B1 (Ascites Free)

clone P2B1, 1 mg/mL, from mouse

Synonym(s):

Platelet endothelial cell adhesion molecule, PECAM-1, EndoCAM, GPIIA′, PECA1, CD31

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

P2B1, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

General description

PECAM-1 is a type I integral membrane glycoprotein, and a member of the immunoglobulin superfamily of cell surface receptors. PECAM-1 is constitutively expressed on the surface of endothelial cells where it is concentrated at cell junctions. PECAM-1 is also expressed at low levels on all leukocytes and platelets. Homotypic interaction of CD31 is important in cell-cell interaction, cell-matrix interaction and signal transduction.

Immunogen

Whole cells expressing human PECAM-1.

Application

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected PECAM-1 in pluripotent cells (Boyd, N. L., et al. (2007). Exp Biol Med. 232(6):833-843.).

Immunoprecipitation Analysis: A representative lot detected PECAM-1 by immunoprecipitation.

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected PECAM-1 in astrocytoma tissue sections (Bronger, H., et al. (2005). Cancer Res. 65(24):11419-28.).

ELISA Analysis: A representative lot of from an independent laboratory detected PECAM-1 in a panel of CD31 mutant cell lines (Newton, J. P, et al. (1997) J. Biol. Chem., 272: 20555-63.).

Immunofluorescence Analysis: A representative lot from an independent laboratory detected PECAM-1 in astrocytoma tissue sections (Bronger, H., et al. (2005). Cancer Res. 65(24):11419-28.).
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-PECAM-1 Antibody, clone P2B1 (Ascites Free) is validated for use in flow cytometry, ICC, IP, IHC, ELISA & immunofluorescence for the detection of PECAM-1.

Quality

Evaluated by Flow Cytometry in human PBMCs.

Flow Cytometry Analysis: 1 µg of this antibody detected PECAM-1 in 1X10E6 human PBMCs.

Target description

~80 kDa calculated

Linkage

Replaces: 04-1074, MAB2148

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laszlo Halasz et al.
International journal of molecular sciences, 25(1) (2024-01-11)
The genome of human adipose-derived stem cells (ADSCs) from abdominal and gluteofemoral adipose tissue depots are maintained in depot-specific stable epigenetic conformations that influence cell-autonomous gene expression patterns and drive unique depot-specific functions. The traditional approach to explore tissue-specific transcriptional
Lei Huang et al.
Nature communications, 8(1), 68-68 (2017-07-14)
Browning of subcutaneous white fat (iWAT) involves several reprograming events, but the underlying mechanisms are incompletely understood. Here we show that the transcription factor Hlx is selectively expressed in brown adipose tissue (BAT) and iWAT, and is translationally upregulated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service