MilliporeSigma
  • Transcription factor Hlx controls a systematic switch from white to brown fat through Prdm16-mediated co-activation.
Transcription factor Hlx controls a systematic switch from white to brown fat through Prdm16-mediated co-activation.
Nature communications (2017-07-14)
Lei Huang, Dongning Pan, Qingbo Chen, Lihua J Zhu, Jianhong Ou, Martin Wabitsch, Yong-Xu Wang
ABSTRACT
Browning of subcutaneous white fat (iWAT) involves several reprograming events, but the underlying mechanisms are incompletely understood. Here we show that the transcription factor Hlx is selectively expressed in brown adipose tissue (BAT) and iWAT, and is translationally upregulated by β3-adrenergic signaling-mediated suppression of the translational inhibitor 4E-BP1. Hlx interacts with and is co-activated by Prdm16 to control BAT-selective gene expression and mitochondrial biogenesis. Hlx heterozygous knockout mice have defects in brown-like adipocyte formation in iWAT, and develop glucose intolerance and high fat-induced hepatic steatosis. Conversely, transgenic expression of Hlx at a physiological level drives a full program of thermogenesis and converts iWAT to brown-like fat, which improves glucose homeostasis and prevents obesity and hepatic steatosis. The adipose remodeling phenotypes are recapitulated by fat-specific injection of Hlx knockdown and overexpression viruses, respectively. Our studies establish Hlx as a powerful regulator for systematic white adipose tissue browning and offer molecular insights into the underlying transcriptional mechanism.The transcriptional co-activator Prdm16 regulates browning of white adipose tissue (WAT). Here, the authors show that Prdm16 interacts with the transcription factor Hlx, which is stabilized in response to β3-adrenergic signaling, to increase thermogenic gene expression and mitochondrial biogenesis in subcutaneous WAT.
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Insulin human, ≥95% (HPLC), semisynthetic, powder, non-sterile
Sigma-Aldrich
apo-Transferrin human, ≥95% protein basis (biuret)
Sigma-Aldrich
Anti-UCP-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PECAM-1 Antibody, clone P2B1 (Ascites Free), clone P2B1, 1 mg/mL, from mouse
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution