Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LSKMTAE50

Millipore

Modified TAE Buffer 50x concentration, 500 mL

Synonym(s):

50 x Modified TAE buffer, Modified TAE Buffer 50x concentration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.25

form

liquid

Quality Level

packaging

pkg of 500 mL

manufacturer/tradename

Montage®

technique(s)

molecular cloning: suitable (BAC prep)
molecular cloning: suitable (plasmid miniprep)

shipped in

ambient

General description

The Montage DNA Gel Extraction Kit provides recovery of 100 to 10,000 bp DNA from agarose gel slices in a single 10-minute spin.

Application

Suitable for use in Montage DNA Gel Extraction Kit

Components

TAE Buffer 50x concentration, 500mL

Physical form

50x conc

Storage and Stability

Reagents should be stored at 15 °C to 30 °C.

Legal Information

MONTAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service