Skip to Content
MilliporeSigma
All Photos(2)

Documents

T9650

Sigma-Aldrich

Tris Acetate-EDTA buffer

10× concentrate, BioReagent, for molecular biology, non-sterile; 0.2 μm filtered

Synonym(s):

TAE buffer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

sterility

non-sterile; 0.2 μm filtered

product line

BioReagent

form

solution

suitability

suitable for gel electrophoresis (after dilution to working concentration)

foreign activity

Protease, none detected
RNAse, none detected

SMILES string

CC(O)=O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3.C2H4O2/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8;1-2(3)4/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2;1H3,(H,3,4)

InChI key

HGEVZDLYZYVYHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tris Acetate-EDTA buffer has been used for the preparation of agarose gel during DNA agarose gel electrophoresis.
TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis. Double-stranded DNA tends to run faster in TAE than in other buffers but can also become exhausted during extended electrophoresis. Buffer circulation or buffer replacement during extended electrophoresis can remedy the lower buffering capacity. Dilution of the concentrated TAE buffer produces a 1× TAE buffer with 40 mM Tris-acetate and 1 mM EDTA, pH 8.3. The 1× TAE buffer is used both in the agarose gel and as a running buffer. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.

Other Notes

0.4 M Tris acetate, pH approx. 8.3, containing 0.01 M EDTA.

Preparation Note

Prepared with Biotechnology Performance Certified Trizma base (T6066) and Molecular Biology Reagent EDTA disodium salt (E5134). Solutions also contain acetic acid (A6283); powdered blend contains Trizma acetate (T1258).
Solution prepared with 18 megohm water

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Circular polymerase extension cloning for high-throughput cloning of complex and combinatorial DNA libraries
Quan J and Tian J
Nature Protocols, 6(2), 242-242 (2011)
Yuksel Temiz et al.
Scientific reports, 8(1), 10603-10603 (2018-07-15)
The ever-increasing need for portable, easy-to-use, cost-effective, and connected point-of-care diagnostics (POCD) has been one of the main drivers of recent research on lab-on-a-chip (LoC) devices. A majority of these devices use microfluidics to manipulate precisely samples and reagents for
Joseph P Torella et al.
Nature protocols, 9(9), 2075-2089 (2014-08-08)
Recombination-based DNA construction methods, such as Gibson assembly, have made it possible to easily and simultaneously assemble multiple DNA parts, and they hold promise for the development and optimization of metabolic pathways and functional genetic circuits. Over time, however, these

Protocols

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service