Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IL029

Sigma-Aldrich

Interleukin-12 Protein, Recombinant human

The recombinant human Interleukin-12 protein (or IL-12 protein) is a potent regulator of cell mediated immune responses & it induces IFN-gamma production by NK & T cells.

Synonym(s):

IL-12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in CHO cells

Assay

>98% (SDS-PAGE and HPLC)

specific activity

≥1.0 x 10E6 U/mg

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 ng/μg endotoxin (of IL-12; 1EU/μg)

input

sample type hematopoietic stem cell(s)

UniProt accession no.

shipped in

dry ice

General description

Interleukin-12 (IL-12) is a potent regulator of cell mediated immune responses and it induces IFN-gamma production by NK and T cells. It is produced by activated monocytes/macrophage cells, B lymphocytes, and connective tissue type mast cells. Among its biological activities, IL-12 promotes the growth and activity of activated NK, CD4+ and CD8+ cells and induces the development of IFN-gamma producing Th1 cells. Human IL-12 is a disulfide linked heterodimeric protein with a molecular weight of 75 kDa comprised of disulfide-bonded 35 kDa (p35) and 40 kDa (p40) subunits.
Product Source: CHO cells (Chinese Hamster Ovarian cells)

Application

For most in vitro applications, IL-12 exerts its biological activity in the concentration range of 1.0 to 10.0 ng/mL.

Biochem/physiol Actions

The ED50 as determined by the induction of IFN-g production from NK cells co-stimulated with IL-18 was ≤ 1.0 ng/mL

Physical form

The sterile filtered solution was lyophilized from 0.5X PBS pH 7.2

Storage and Stability

The lyophilized IL-12, though stable at room temperature, is best stored desiccated below 0°C. Reconstitute with sterile water to a concentration of 0.2 mg/mL. Reconstituted IL-12 should be stored in undiluted aliquots at -20°C for up to 6 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiyun Kim et al.
Biomicrofluidics, 9(4), 044109-044109 (2015-09-05)
Fabrication methods for the development of multiplexed immunoassay platforms primarily depend on the individual functionalization of reaction chambers to achieve a heterogeneous reacting substrate composition, which increases the overall manufacturing time and cost. Here, we describe a new type of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service