Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H7666

Sigma-Aldrich

IL-12 human

HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture

Synonym(s):

Interleukin-12 human, IL-12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Interleukin-12 human, HumanKine®, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin, tested

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

≤2 ng/mL EC50

mol wt

dimer 57 kDa (glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Interleukin-12 (IL-12) human has been used to study IL-12 mediated enhancement of IL-12Rβ2 (IL-12 receptor) expression.

Biochem/physiol Actions

Interleukin-12 (IL-12) was identified as a factor secreted by human Epstein-Barr (EBV)-transformed B cell lines. IL-12, or Natural Killer Cell Stimulatory Factor (NKSF), is a 75 kDa disulfide-linked heterodimer of a 35 kDa subunit and 40 kDa subunit. IL-12 is produced predominantly by monocytes and NK cells and induces T cells and NK cells to produce IFN-γ. Human IL-12 is not active on mouse cells, but murine IL-12 is active on both murine and human lymphocytes. The biological activity of recombinant human IL-12 was measured by its ability to stimulate the proliferation of PHA-activated human T lymphoblasts. IL-12 has anti-angiogenic properties.

Physical form

lyophilized from a 0.2 μm filtered solution of PBS

Analysis Note

The specific activity was determined by the dose-dependent release of IFN-γ from the human NK92 cell line in presence of 20 ng/mL rIL-2.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

T-bet upregulation and subsequent interleukin 12 stimulation are essential for induction of Th1 mediated immunopathology in Crohn's disease.
Matsuoka K et al.
Gut, 53, 1303-1303 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service