Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ES-003

Sigma-Aldrich

EmbryoMax® Electroporation Buffer

The EmbryoMax Electroporation buffer is used to dissolve the DNA that is to be transfected into mouse embryonic stem cells.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122100
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

liquid

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type: mouse embryonic stem cell(s)
sample type induced pluripotent stem cell(s)

General description

This Electroporation buffer is used to dissolve the DNA that is to be transfected into the ES cells. Generally 25-40μg of the knockout construct DNA is dissolved in 30μl of the buffer.

Storage and Stability

Store at 2-8°C upto 18 months.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture

Protocols

Stem Cell protocols for cryopreservation, thawing of cryopreserved stem cells and media preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service