Skip to Content
MilliporeSigma

ES-006

EmbryoMax® Gelatin Solution

0.1% Gelatin Solution is available in a 500 mL format and may be used for routine mouse embryonic stem cell culture applications.

Synonym(s):

Porcine Gelatin Coating Solution, porcine gelatin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

EmbryoMax® 0.1% Gelatin Solution, The EmbryoMax 0.1% Gelatin Solution is available in a 500 mL format and may be used for routine mouse embryonic stem cell culture applications.

Quality Segment

form

liquid

manufacturer/tradename

Specialty Media, EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)

Application

Procedure for Coating Multiwell Dishes

1. Add the appropriate amounts of 0.1% Gelatin in Sterile Water to each well of the plate as indicated in the table below:

Wells_Amount/Well
96100 μL
48300 μL
240.5 mL
121.0 mL
61.5-2.0 mL

2. Leave the gelatin solution on the wells for at least 30 minutes at room temperature, with dish lids on.

3. Remove the gelatin solution and discard.

4. Immediately add cells to the dish.

Note: Do not allow the dishes to dry before adding cells.

Physical form

EmbryoMax 0.1% Gelatin Solution. Gelatin is sourced from porcine. Product is ready to use for plating.

Preparation Note

Store at ambient temperature for upto 2 years

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

The Luminata ELISA HRP chemiluminescent substrates provide three major advantages: 1. High sensitivity for detecting as little as 0.1 pg of protein. 2. Broad dynamic detection range (up to 2,500-fold), enabling a wide range of quantitation. 3. Flexibility to adjust signal strength by one of two methods: a. Exposing the well to the luminometer for a longer time. b. Applying a more sensitive substrate to obtain signals in wells with otherwise undetectable signals.

View All Related Content





Global Trade Item Number

SKUGTIN
ES-006-B08436037123900
ES-006-BK04053252903519