Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

BAH136800040

Flowmi® Cell Strainers

porosity 40 μm, for 1000 uL Pipette Tips, pack of 50 ea

Synonym(s):

compact laboratory sample filters, facs sample filtration, small sample filter, small sample pipette tip filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104932
NACRES:
NB.01

material

white

sterility

sterile

packaging

pack of 50 ea
pkg of (supplied in a tray)

manufacturer/tradename

Bel-Art H13680-0040

pore size

40 μm porosity

Looking for similar products? Visit Product Comparison Guide

General description

Scienceware® Flowmi Cell Strainers provide fast and efficient filtering of small volume samples (up to 1000μl) prior to FLOW or Analysis. Patent pending.

Features and Benefits

  • Preserves volume when filtering small sample volumes
  • Decreases the potential for clogging of FLOW or FACS
  • instruments by effectively filtering cellular debris
  • Use with your favorite pipette tips; Flowmi fits most 1000μl tips including Fisherbrand Sure-One, VWR brand, Axygen, Nichiryo and Eppendorf and more
  • Compact tray holds 50 Flowmi Cell Strainers ready for press and fit attachment to your tip; features a sliding cover for easy, one-handed use
  • Packaged in a re-sealable bag to maintain sterility when used in laminar flow hoods

Other Notes

Recommended for use with samples having a maximum concentration of 2MM cells/ml.

Legal Information

Flowmi is a registered trademark of SP Scienceware, Division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zong Miao et al.
PLoS genetics, 16(9), e1009018-e1009018 (2020-09-15)
Reverse causality has made it difficult to establish the causal directions between obesity and prediabetes and obesity and insulin resistance. To disentangle whether obesity causally drives prediabetes and insulin resistance already in non-diabetic individuals, we utilized the UK Biobank and
Marcus Alvarez et al.
Scientific reports, 10(1), 11019-11019 (2020-07-06)
Single-nucleus RNA sequencing (snRNA-seq) measures gene expression in individual nuclei instead of cells, allowing for unbiased cell type characterization in solid tissues. We observe that snRNA-seq is commonly subject to contamination by high amounts of ambient RNA, which can lead
Jeremy Lotto et al.
Cell, 183(3), 702-716 (2020-10-31)
The cellular complexity and scale of the early liver have constrained analyses examining its emergence during organogenesis. To circumvent these issues, we analyzed 45,334 single-cell transcriptomes from embryonic day (E)7.5, when endoderm progenitors are specified, to E10.5 liver, when liver
Pavlo A Nesterenko et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(3) (2021-01-13)
T cell receptors (TCRs) are generated by somatic recombination of V/D/J segments to produce up to 1015 unique sequences. Highly sensitive and specific techniques are required to isolate and identify the rare TCR sequences that respond to antigens of interest.

Articles

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service