Skip to Content
MilliporeSigma
All Photos(1)

Documents

BI0830

Sigma-Aldrich

Deblocking Reagent 3% Trichloroacetic Acid in Dichloromethane

Novabiochem®

Synonym(s):

DNA Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006

product line

Novabiochem®

Quality Level

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: Lewis acid

manufacturer/tradename

Novabiochem®

shipped in

ambient

storage temp.

2-30°C

General description

Biosynthesis Novabiochem®

Application

BI0830-0450 is packaged in 28/405 glass bottles
BI0830-0950 is packaged in 38/430 glass bottles
BI0830-4000 is packaged in 38/430 glass bottles
BI0830-P4000 is packaged in 38/430 poly coated glass bottles
DNA Reagents Label Change Explanation

Analysis Note

Color (APHA): 20APHA max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Trichloroacetic acid (by titration): 2.95-3.05% w/v
Water (H2O): 200ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service