Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX0151

Supelco

Acetonitrile

≥99.9% (GC), anhydrous, Biosynthesis, OmniSolv®

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12352117
EC Index Number:
200-835-2
grade:
anhydrous
technique(s):
GC/GC: suitable
application(s):
pharmaceutical
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

Product Name

Acetonitrile, anhydrous, Biosynthesis OmniSolv®, ≥99.9% (GC)

grade

anhydrous

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

description

gradient analysis: <=0.003 at 210nm

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

GC/GC: suitable

impurities

≤0.16 μeq/g Titrable base
≤0.20 ppm Free amine (as dimethylamine)
≤0.8 μeq/g Titrable acid
≤10 ppm Water (H2O)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

UV cutoff

≤189 nm

UV absorption

λ: 189 nm Amax: ≤1.00
λ: 195 nm Amax: ≤0.15
λ: 210 nm Amax: ≤0.03
λ: 230 nm Amax: ≤0.005
λ: 250 nm Amax: ≤0.005
λ: 270 nm Amax: ≤0.005

application(s)

pharmaceutical

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

AX0151P-1 is packaged in poly-coated glass bottles.
anhydrous, Biosynthesis OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)250ppt maxFormClear liquidFree amine (as dimethylamine)0.20ppm maxGradient at 210 nm0.003AU maxInfrared SpectrumConforms to standardRefractive index (at 20°C)1.343-1.345Titrable acid0.8µeq/g maxTitrable base0.16µeq/g maxUV Abs. at 189 nm1.00AU maxUV Abs. at 195 nm0.15AU maxUV Abs. at 210 nm0.03AU maxUV Abs. at 230 nm0.005AU maxUV Abs. at 250 nm0.005AU maxUV Abs. at 270 nm0.005AU maxUV Cut-off189nm maxWater (H2O)10ppm max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service