Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ACK5030NT

Millipore

Amicon® Pro Affinity Concentration Kit Ni-NTA with 30kDa Amicon® Ultra-0.5 Device

Amicon Pro Affinity Concentration Kit Ni-NTA - A Centrifugal Tool for purifying & concentrating His-tagged proteins.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101

material

Amicon Ultra styrene-butadiene device
Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding)
polyethylene cap
polyethylene frit
polypropylene device (for exchange)
polypropylene frit
polypropylene tube (AU0.5; for collection)
polypropylene tube (for collection)
polypropylene tube (holder)

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Pro

parameter

10 mL sample volume

technique(s)

buffer exchange: suitable
protein purification: suitable (antibody labeling)
protein purification: suitable (concentration)
protein purification: suitable (enrichment)

L

12 cm (4.7 in.)

diam.

2.97 cm

filtration area

1 cm2

working volume

10 mL

pore size

30 kDa NMWCO

shipped in

wet ice

General description

Filter Code: PLCGC
Filter Type: Ultrafiltration
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO 30 kDa, 3 ml HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25ml Bind Buffer (4X), 25 ml Wash buffer (4X), 5ml elution buffer (4X)

Application

Amicon Pro Affinity Concentration Kit Ni-NTA - A Centrifugal Tool for purifying & concentrating His-tagged proteins.

Features and Benefits

Minimum Final Concentrate Volume: 15 µL

Physical form

Color Code: Clear with pink cap

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

Flash Point(F)

95.9 °F

Flash Point(C)

35.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service