Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC1469

Sigma-Aldrich

Anti-NRP1

from rabbit, purified by affinity chromatography

Synonym(s):

Vascular endothelial cell growth factor 165 receptor, CD304

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... NRP1(8829)

General description

Neuropilin-1 (UniProt: O14786; also known as Vascular endothelial cell growth factor 165 receptor, CD304) is encoded by the NRP1 (also known as NRP, VEGF165R) gene (Gene ID: 8829) in human. Three isoforms of Neuropilin-1 have been reported that are produced by alternative splicing. Isoform 1 of Neuropilin-1 is a homodimer, single-pass type I membrane protein with three calcium-binding sites (aa 195, 209, and 250). It can also form heterodimers with neuropilin-2. This membrane-bound isoform serves as a receptor involved in the development of the cardiovascular system, in angiogenesis, in the formation of certain neuronal circuits and in organogenesis outside the nervous system. It mediates the chemorepulsant activity of semaphorins. It binds to semaphorin 3A, The PLGF-2 isoform of PGF, The VEGF165 isoform of VEGFA and VEGFB. Its co-expression with VEGFR-2 (KDR) results in increased VEGF165 binding to VEGFR-2 as well as increased chemotaxis. It has an extracellular domain (aa 22-856), a helical domain (aa 857-879), and the cytoplasmic domain (aa 880-923). It is produced with a signal peptide (aa 1-21) that is subsequently cleaved to generate the mature form. The soluble isoform 2 binds VEGF-165 and appears to inhibit its binding to cells. It may also induce apoptosis by sequestering VEGF-165. It may also bind various members of the semaphorin family and its expression has an adverse effect on blood vessel number and integrity. The expression of isoforms 1 and 2 does not seem to overlap. Isoform 1 is expressed by the blood vessels of different tissues. In the developing embryo it is found predominantly in the nervous system. In adult tissues, it is highly expressed in heart and placenta; moderately in lung, liver, skeletal muscle, kidney and pancreas; and low in adult brain. Isoform 2 is found in liver hepatocytes, kidney distal and proximal tubules.

Specificity

This rabbit polyclonal antibody detects Neuropilin-1 in human cells. It targets an epitope within 313 amino acids from the internal region.

Immunogen

His-tagged recombinant fragment corresponding to 313 amino acids from the internal region of human Neuropilin-1.

Application

Anti-NRP1, Cat. No. ABC1469, is a rabbit polyclonal antibody that detects human Neuropilin-1 and has been tested for use in Immunohistochemistry (Paraffin), Inhibition Activity/Function, and Western Blotting.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected NRP1 in human prostate cancer and human bone marrow tissue.

Western Blotting Analysis: A representative lot detected NRP1 in Western Blotting applications (Pang, H.B., et. al. (2014). Nat Commun. 5:4904).

Inhibits Activity/Function: A representative lot reduced iRGD-induced vascular permeability in tumor tissue and reduced the uptake of nanoparticles. (Pang, H.B., et. al. (2015). Sci Adv. 1(10):e1500821; Sugahara, K.N., et. al. (2010). Science. 328(5981):1031-5).

Quality

Evaluated by Western Blotting with recombinant Neuropilin-1.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected recombinant Neuropilin-1. recombinant protein.

Target description

~36 kDa observed for recombinant fragment. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service