Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-085

Supelco

(±)-Propranolol-D7 (ring-D7) solution

100 μg/mL in methanol with 5% 1 M HCl, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Propranolol-d7 (ring-d7)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14D7NO2
CAS Number:
Molecular Weight:
266.39
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol with 5% 1 M HCl

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

N(C(C)C)CC(O)COc1c2c(c(c(c1[2H])[2H])[2H])c(c(c(c2[2H])[2H])[2H])[2H]

InChI

1S/C16H21NO2/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16/h3-9,12,14,17-18H,10-11H2,1-2H3/i3D,4D,5D,6D,7D,8D,9D

InChI key

AQHHHDLHHXJYJD-MIBSYSDMSA-N

General description

Propranolol is a sympatholytic, non-selective beta blocker used to treat hypertension, anxiety and panic. The drug is sold under numerous brand names such as Inderal®, Avlocardyl, Deralin, and Dociton. This internal standard is suitable for quantitation of propranolol levels in patient samples using LC/MS or GC/MS for applications in clinical toxicology, diagnostic testing, forensic analysis, pharmaceutical research and isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Inderal is a registered trademark of Cranford Pharmaceuticals, LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service