Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5603-AF647

Sigma-Aldrich

Anti-Sox2 Antibody, Alexa Fluor 647 Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

conjugate

ALEXA FLUOR 555

Quality Level

clone

polyclonal

purified by

affinity chromatography

species reactivity

rabbit, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable

UniProt accession no.

General description

KLH-conjugated linear peptide corresponding to a C-terminal region sequence of human Sox2.

Application

Epitope: C-terminal region.

Quality

Evaluated by Immunocytochemistry in mouse ESCs.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Sox2 in mouse ESCs.

Physical properties

~39 kDa observed.

Physical form

Purified rabbit polyclonal antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service