Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE992

Sigma-Aldrich

Anti-Septin-9 Antibody, clone 10C10

clone 10C10, from mouse

Synonym(s):

Septin-9, MLL septin-like fusion protein, MLL septin-like fusion protein MSF-A, Ovarian/Breast septin, Ov/Br septin, SeptD1, Septin D1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10C10, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SEPTIN9(10801)

General description

Septin-9 (UniProt Q9UHD8; also known as MLL septin-like fusion protein, MLL septin-like fusion protein MSF-A, Ovarian/Breast septin, Ov/Br septin, SeptD1, Septin D1) is encoded by the SEPT9 (also known as AF17q25, KIAA0991, MSF, MSF1, PNUTL4, SINT1) gene (Gene ID 10801) in human. Septins are GTPases capable of forming higher-order oligomers and filamentous polymers that contact with cell membrane and cytoskeleton. Septins are involved in various cellular processes, including cytokinesis, karyokinesis, exocytosis, and cell shape maintenance. Septins exist in complexes that are composed of ordered non-polar arrays of multiple different septins. Human genome contains 13 septin genes that are grouped into the SEPT2 subgroup (septins 1, 2, 4, and 5), the SEPT6 subgroup (septins 6, 8, 10, 11, and 14), the SEPT3 subgroup (septins 3, 9, and 12), and the SEPT7 subgroup (septin 7). Among mammalian septins, septin-9 undergoes the most alternative splicings, alteration in septin-9 isoforms expression has been linked to cancer and SEPT9 gene mutations have been linked to hereditary neuralgic amyotrophy. Cdk1-catalyzed septin-9 Thr24 phosphorylation is reported to promote septin-9 interaction with the peptidylprolyl isomerase Pin1. Genetic knockdown of either protein results in abscission defect during cell division.

Specificity

Clone 10C10 specifically detected purified recombinant human Septin-9, but not recombinant Spetin-1, -2, -5, -6, -7, -8. -9. -11, or -12, by Western blotting (Estey, M.P., et al. (2010). J. Cell Biol. 191(4):741-749). The epitope has been mapped to region present in human Septin-9 spliced isoforms 1 (Epsilon, MSF-A), 2 (Alpha), 5 (Gamma), and 7, but absent in spliced isoforms 3 (Beta, MSF-B), 4 (Delta), 8, and 9 reported by UniProt (Q9UHD8).

Immunogen

Epitope: Internal (N-terminal half).
His-tagged recombinant human septin-9.

Application

Immunocytochemistry Analysis: A representative lot detected septin-9 immunoreactivity in U2OS cells (Courtesy of Dr. William S. Trimble, University of Toronto, Canada).
Immunocytochemistry Analysis: Clone 10C10 hybridoma culture supernatant immunostained intracellular septin-9 in hTERT-immortalized retinal pigment epithelial (hTERT-RPE) cells (Courtesy of Dr. William S. Trimble, University of Toronto, Canada).
Immunocytochemistry Analysis: A representative lot detected septin-9 immunoreactivity colocalized with septin-2 and septin-11 at the cleavage furrow in anaphase and the midbody in telophase HeLa cells (Estey, M.P., et al. (2010). J. Cell Biol. 191(4):741-749).
Immunoprecipitation Analysis: A representative lot immunoprecipited septin-9 from mitotically arrested HeLa cells following nocodazole (Cat. No. 487928) treatment. Cdk1 inhibition by Roscovitine (Cat. No. 57360) treatment or siRNA-meidated Cdk1 knowndown diminished the phosphroylation of the immunoprecipitated septin-9 (Estey, M.P., et al. (2013). J. Biol. Chem. 288(42):30075-30086).
Immunoprecipitation Analysis: A representative lot immunoprecipitated significantly more septin-9 from unsynchronized than from mitotically synchronized HeLa cells, the difference was normalized following siRNA-mediated Pin1 knockdown (Estey, M.P., et al. (2013). J. Biol. Chem. 288(42):30075-30086).
Western Blotting Analysis: A representative lot specifically detected recombinant septin-9 spliced isoforms and fragments that contain the epitope region sequence, but not recombinant isoforms or fragments lacking the epitope (Estey, M.P., et al. (2013). J. Biol. Chem. 288(42):30075-30086).
Western Blotting Analysis: A representative lot specifically detected purified recombinant human septin-9, but not purified recombinant spetin-1, -2, -5, -6, -7, -8. -9. -11, -12 (Estey, M.P., et al. (2010). J. Cell Biol. 191(4):741-749).
Western Blotting Analysis: A representative lot detected the presence of co-precipitated septin-9 in septin-2 immunoprecipitates from HeLa cell lysates (Estey, M.P., et al. (2010). J. Cell Biol. 191(4):741-749).
Note: For Western blotting application, prior enrichment of septin-9 by immunoprecipitation or other purification methods is highly recommended. Clone 10C10 is not recommended for probing septin-9 using whole cell lysates.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
This Anti-Septin-9 Antibody, clone 10C10 is validated for use in Western Blotting, Immunocytochemistry, Immunoprecipitation for the detection of Septin-9.

Quality

Evaluated by Western Blotting of recombinant human septin-9.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected His-tagged recombinant human septin-9.

Target description

~69 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilona A Kesisova et al.
The Journal of cell biology, 220(2) (2021-01-09)
The metabolic and signaling functions of lysosomes depend on their intracellular positioning and trafficking, but the underlying mechanisms are little understood. Here, we have discovered a novel septin GTPase-based mechanism for retrograde lysosome transport. We found that septin 9 (SEPT9)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service