Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
SKB1, monoclonal
Application:
FACS, ICC, IP, WB
Citations:
65
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
SKB1, monoclonal
species reactivity
rat, human, mouse
packaging
antibody small pack of 25 μg
manufacturer/tradename
Upstate®
technique(s)
flow cytometry: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
mouse ... Akt1(11651)
General description
Akt (protein kinase B), a serine/threonine kinase, has emerged as a critical enzyme in signal transduction pathways involved in cell proliferation, apoptosis, angiogenesis, and diabetes. In mammals three isoforms of Akt (α, β, γ or Akt 1, 2, 3) are reported that exhibit a high degree of homology, but differ slightly in the localization of their regulatory phosphorylation sites. Akt α is the predominant isoform in most tissues, whereas the highest expression of Akt β is observed in the insulin-responsive tissues, and Akt γ is abundant in brain tissue. Each Akt isoform is composed of three functionally distinct regions: an N-terminal pleckstrin homology (PH) domain that provides a lipid-binding module to direct Akt to PIP2 and PIP3, a central catalytic domain, and a C-terminal hydrophobic motif.
60 kDa
Application
Detect Akt/PKB using this Anti-Akt/PKB Antibody, PH Domain, clone SKB1 validated for use in FC, IC, IP & WB.
Physical form
Format: Purified
Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.02 M phosphate buffer, pH 7.6, 0.25M NaCl, 0.1% sodium azide and 30% glycerol.
Analysis Note
Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.
routinely evaluated by immunoblot on RIPA lysates from A431 cells
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Evaluation of approaches to generation of tissue-specific knock-in mice.
Bayascas, JR; Sakamoto, K; Armit, L; Arthur, JS; Alessi, DR
The Journal of Biological Chemistry null
Heng-Liang Lin et al.
The Journal of pharmacology and experimental therapeutics, 323(1), 365-373 (2007-07-25)
Combretastatin A4 (CA4) is a drug that targets tumor vasculature to inhibit angiogenesis. Whether CA4 has a direct effect on gastric cancer is not known. We herein investigated the effect of CA4 on growth and metastasis of gastric cancer cells
HRT1 modulates vascular smooth muscle cell proliferation and apoptosis.
Wenli Wang, Chengyu Z Prince, Xing Hu, Matthew J Pollman
Biochemical and biophysical research communications null
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 05-591-25UG | 04054839337703 |