Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5583

Sigma-Aldrich

Anti-Nerve Growth Factor Antibody, pro

Chemicon®, from rabbit

Synonym(s):

NGF, pro

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NTF3(4908)

Specificity

Recognizes proNGF (Nerve Growth Factor).

SPECIES REACTIVITIES:

The immunogen sequence has 100% homology with mouse and 76% homology with human proNGF. Reactivity with other species has not yet been tested.

Immunogen

Synthetic corresponding to amino acids 84-104 of rat proNGF (Accession P25427).

Application

Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins
This Anti-Nerve Growth Factor Antibody, pro is validated for use in WB, IH for the detection of Nerve Growth Factor.
Western blotting: 1:200-1:500 using ECL on His-fusion protein containing the entire prodomain of rat proNGF.

Immunohistochemistry on rat brain sections.

Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.025% sodium azide. Reconstitute with 50 mL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months after date of receipt. After reconstitution maintain antibody and control peptide at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile distilled water. For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wohaib Hasan et al.
Autonomic neuroscience : basic & clinical, 145(1-2), 17-26 (2008-11-21)
Cardiac function is regulated by interactions among intrinsic and extrinsic autonomic neurons, and the mechanisms responsible for organizing these circuits are poorly understood. Parasympathetic neurons elsewhere synthesize the neurotrophin NGF, which may promote postganglionic axonal associations where parasympathetic axons inhibit
María Eugenia Bogetti et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 71, 111-121 (2018-08-31)
A decrease in the concentration of oxygen in the blood and tissues (hypoxia) produces important, sometimes irreversible, damages in the central nervous system (CNS) both during development and also postnatally. The present work aims at analyzing the expression of nerve
F Jardí et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 24(12), e580-e590 (2012-10-18)
Nerve growth factor (NGF)-mucosal mast cell (MMC) interaction has been implicated in the remodeling of enteric circuitries and associated functional changes. We investigated the involvement of NGF and its receptor TrkA in the altered colonic contractile activity observed in the
Zheng-Mao Li et al.
MedComm, 3(3), e128-e128 (2022-07-01)
Accumulating evidence indicates that epilepsy has a higher risk of inducing memory impairment and dementia. However, the underlying onset mechanism remains unclear. Here, we found that mice with spontaneous epilepsy induced by endothelial CDK5 deficiency exhibited hippocampal-dependent memory impairment at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service