Skip to Content
MilliporeSigma
All Photos(5)

Documents

AB5475

Sigma-Aldrich

Anti-Prox 1 Antibody

serum, Chemicon®

Synonym(s):

Anti-A230003G05Rik, Anti-PROX-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, zebrafish

species reactivity (predicted by homology)

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PROX1(5629)

Related Categories

General description

Prox1, the homolog of Drosophila Prospero, is a homeobox-containing transcription factor that binds and functions as a co-receptor of liver receptor homolog 1 (LRH1/NR5A2). It is a specific marker for lymphatic endothelial cells, as well as hepatoblasts and hepatocytes. Human Prox1 shares 97% amino acid sequence identity with mouse and rat Prox1.

Specificity

Other species have not been tested.
Recognizes Prox1. The antibody is useful for identifying Prox1 expressing cells during the development of many areas of the nervous system including the dorsal spinal cord, ganglionic eminences, thalamus and hippocampus.

Immunogen

Synthetic peptide from the C-terminus of mouse Prox1.

Application

Detect Prox 1 using this Anti-Prox 1 Antibody validated for use in IC, IH.
Immunohistochemistry:
1:2,000-1:10,000 with overnight incubation on 4% paraformaldehyde fixed, frozen tissue sections. For embryos immersion fixation is recommended. For older animals perfusion fixation is recommended. Suggested permeabilization solution is 0.1% Triton X100. Suggested blocking buffer is PBS with 0.1% Triton and 10% serum.

Immunocytochemistry:
A previous lot of this antibody was used on Immunocytochemistry

Physical form

Rabbit serum in 50 % glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Embryonic brain tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ubiquitin proteasome system is required for cell proliferation of the lens epithelium and for differentiation of lens fiber cells in zebrafish.
Imai, F; Yoshizawa, A; Fujimori-Tonou, N; Kawakami, K; Masai, I
Development null
Decrease in tonic inhibition contributes to increase in dentate semilunar granule cell excitability after brain injury.
Gupta, A; Elgammal, FS; Proddutur, A; Shah, S; Santhakumar, V
The Journal of Neuroscience null
Notum homolog plays a novel role in primary motor innervation.
Cantu, JA; Flowers, GP; Topczewski, J
The Journal of Neuroscience null
Granule cells in the CA3 area.
Szabadics, J; Varga, C; Brunner, J; Chen, K; Soltesz, I
The Journal of Neuroscience null
Natalia G Kan et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(10), 3516-3525 (2009-06-24)
Here, we describe the zebrafish (Danio rerio) as a vertebrate model system to study liver regeneration with the added benefit of its powerful genetics and screening possibilities to uncover the molecular pathways underlying liver regeneration. We developed a partial hepatectomy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service