Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

7.4.C4

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 92092321, Reactivity: Anti-Fish Leukocytes, red blood cells and endothelial cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 92092321-DNA-5UG
pkg of vial of cells 92092321-1VL

ImmunoDon

(C57/BL/6) mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-Fish Leukocytes, red blood cells and endothelial cells

technique(s)

cell culture | mammalian: suitable

isotype

IgG1, kappa

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse (C57/BL/6) spleen

Cell Line Description

The antibody is directed against cell surface components expressed on leukocytes, red blood cells and vascular endothelial cells of fish.

Immunogen

Optic tectum from optic nerves of Oreochromis mossambicus 12 days post injury

Application

Immunohistochemistry

Culture Medium

DMEM + 2mM glutamine + 1% Hypoxanthine, Thymidine (HT) supplement + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 8% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service