Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3506P

Sigma-Aldrich

Anti-Arginine Vasopressin Receptor 1a Antibody

Chemicon®, from rabbit

Synonym(s):

AVP R1a, V1a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes rat Arginine Vasopressin Receptor V1a (AVR-V1a). The immunogen shows no significant sequence homology with AVR-V1b, AVR-V2 or AVR-V3.

SPECIES REACTIVITIES: The immunogen peptide is 84% conserved in mouse, 68% in human and 52% in sheep AVR-V1a. Reactivity with other species has not been confirmed.

Immunogen

A 19 amino acid peptide sequence within the extracellular, N-terminal domain of rat AVR-V1a (Morel et al. 1992; Innamorti et al. 1996; Thibonnier et al. 1994; Hirasawa et al. 1994).

Application

Anti-Arginine Vasopressin Receptor 1a Antibody detects level of Arginine Vasopressin Receptor 1a & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors
Western blot: 1-10 μg/mL using ECL.

ELISA: 0.5-1 μg/mL using 1 μg/mL control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol A Dannenhoffer et al.
Psychopharmacology, 235(10), 3065-3077 (2018-08-25)
Adolescent intermittent ethanol exposure (AIE) produces lasting, sex-specific social anxiety-like alterations in male, but not female rats. Oxytocin (OXT) and vasopressin (AVP) brain systems play opposite roles in regulating social preference/avoidance, with OXT increasing approach to, and AVP increasing avoidance
Elie Simard et al.
PloS one, 10(8), e0128881-e0128881 (2015-08-08)
Increased blood glucose concentrations promote reactions between glucose and proteins to form advanced glycation end-products (AGE). Circulating AGE in the blood plasma can activate the receptor for advanced end-products (RAGE), which is present on both endothelial and vascular smooth muscle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service