Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2172

Sigma-Aldrich

CLP290

≥98% (HPLC)

Synonym(s):

[5-Fluoro-2-[(Z)-(2-hexahydropyridazin-1-yl-4-oxo-thiazol-5-ylidene)methyl]phenyl] pyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21FN4O3S
CAS Number:
Molecular Weight:
404.46
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C(/C=C2C(N=C(N3NCCCC3)S/2)=O)C(OC(N4CCCC4)=O)=C1

InChI

1S/C19H21FN4O3S/c20-14-6-5-13(15(12-14)27-19(26)23-8-3-4-9-23)11-16-17(25)22-18(28-16)24-10-2-1-7-21-24/h5-6,11-12,21H,1-4,7-10H2/b16-11-

InChI key

DIXMMXNNKLCLOM-WJDWOHSUSA-N

Biochem/physiol Actions

CLP290 is a small molecule enhancer of KCC2 activity. It is an orally active and more bioavailable precursor of the selective K+-Cl- cotransporter KCC2 activator CLP257. CLP290 was recently shown to restore Cl- transport and rescue morphine-induced hyperalgesia (MIH) in morphine-treated rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Gagnon et al.
Nature medicine, 19(11), 1524-1528 (2013-10-08)
The K(+)-Cl(-) cotransporter KCC2 is responsible for maintaining low Cl(-) concentration in neurons of the central nervous system (CNS), which is essential for postsynaptic inhibition through GABA(A) and glycine receptors. Although no CNS disorders have been associated with KCC2 mutations
Francesco Ferrini et al.
Scientific reports, 7(1), 3870-3870 (2017-06-22)
Morphine-induced hyperalgesia (MIH) is a severe adverse effect accompanying repeated morphine treatment, causing a paradoxical decrease in nociceptive threshold. Previous reports associated MIH with a decreased expression of the Cl
Akiko Doi et al.
PloS one, 16(3), e0248113-e0248113 (2021-03-13)
Immature neurons dominantly express the Na+-K+-2Cl- cotransporter isoform 1 (NKCC1) rather than the K+-Cl- cotransporter isoform 2 (KCC2). The intracellular chloride ion concentration ([Cl-]i) is higher in immature neurons than in mature neurons; therefore, γ-aminobutyric acid type A (GABAA) receptor
Ming Chen et al.
eLife, 6 (2017-01-06)
Amyloid precursor protein (APP) is enriched at the synapse, but its synaptic function is still poorly understood. We previously showed that GABAergic short-term plasticity is impaired in App knock-out (App-/-) animals, but the precise mechanism by which APP regulates GABAergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service