Key Documents
OH30031656
Ohaus® Dissolved Oxygen Meter
Model ST300D-B
About This Item
UNSPSC Code:
41115611
material
epoxy shaft
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30031656
dissolved oxygen measuring range
0.0-199.9%
temperature measuring range
0-50 °C
battery power
4 AAA, 250 hr life
General description
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results.
Application
Dissolved oxygen (DO) measurement
Field and lab Dissolved Oxygen measurement
Features and Benefits
30 measurement memory
99 measurement memory, last calibration data recall, auto barometric pressure measurement, auto temperature compensation with manual salinity compensation
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)