Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1787P

Sigma-Aldrich

Anti-Dopamine D4 Receptor Antibody, extracellular domain

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Drd4(13491)

Specificity

Specific for Dopamine Receptor D4. Mouse Dopamine receptor D4 is a 387 aa, G-protein coupled and transmembrane receptor protein (Van Tol et al. 1992; Legerski & Peterson 1992; Livingstone et al. 1992; Seeman et al. 1994). The immunogen peptide shows no significant homology with other dopamine receptors (D1, D2, D3 or D5).

SPECIES REACTIVITIES: The immunogen peptide shows sequence homology of 90% with rat and human D4 receptors (Van Tol et al. 1992; Legerski & Peterson 1992; Livingstone et al. 1992; Seeman et al. 1994; O′Malley et al. 1992). Antibody cross-reactivity with D4R from various species is not known.

Immunogen

A 10 amino acid peptide sequence from the mouse D4 receptor within the 3rd extracellular domain.
Epitope: extracellular domain

Application

Anti-Dopamine D4 Receptor Antibody, extracellular domain is an antibody against Dopamine D4 Receptor for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1-10 μg/mL (Chemiluminescence technique). Approximately 100-400 μg of membrane protein/lane should be loaded to visualize the receptor band. Antibody detects ~55kDa band in HT222 cells and mouse hippocampal membranes. {http://www.jneurosci.org/cgi/reprint/21/16/6069.pdf, Ishige, K et.al.(2001) J. Neuroscience 21(16):6069-6076.

ELISA: 0.5-1.0 μg/mL (1 μg/mL immunogen peptide/well)

Not tested in immunohistochemical methods.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB1789P

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphane F Maison et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(1), 344-355 (2012-01-06)
Pharmacological studies suggest that dopamine release from lateral olivocochlear efferent neurons suppresses spontaneous and sound-evoked activity in cochlear nerve fibers and helps control noise-induced excitotoxicity; however, the literature on cochlear expression and localization of dopamine receptors is contradictory. To better

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service