Skip to Content
MilliporeSigma
All Photos(1)

Documents

AAWG04700

Millipore

MCE Membrane Filter, 0.8 μm Pore Size

MF-Millipore, filter diam. 47 mm, hydrophilic, white, gridded

Synonym(s):

MF-Millipore Membrane Filter, 0.8 µm pore size, gridded

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

gridded
mixed cellulose esters (MCE) membrane
white filter

Quality Level

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, gridded, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

16 L/min-cm2 air flow rate
190 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

47 mm

thickness

150 μm

gravimetric extractables

4%

color

white

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.8 μm pore size
82 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥1.0 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: AAWG

Application

  • Air monitoring
  • Particle monitoring
  • Particle removal
  • Bioassays
  • Microplastics

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service