Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAWG04700

Millipore

MCE Membrane Filter, 1.2 μm Pore Size

MF-Millipore, filter diam. 47 mm, hydrophilic, white, gridded

Synonym(s):

MF-Millipore Membrane Filter, 1.2 µm pore size, gridded

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

gridded
mixed cellulose esters (MCE) membrane
white filter

Quality Level

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, gridded, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

20 L/min-cm2 air flow rate
270 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

47 mm

thickness

150 μm

gravimetric extractables

5%

color

white

refractive index

n/D 1.512

matrix

MF-Millipore

pore size

1.2 μm pore size
82 % porosity

bubble point

≥0.76 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Filter Code: RAWG

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha M Peneyra et al.
Zebrafish, 15(1), 63-72 (2017-10-20)
Pseudoloma neurophilia, a microsporidium that primarily infects neural tissues, is a common pathogen in laboratory zebrafish. The risk of parasite transmission with different spawning apparatuses and the effectiveness of disinfection are unknown. In this study, we spawned uninfected zebrafish with

Articles

An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.

An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.

An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.

An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service