Skip to Content
MilliporeSigma

SMWP04700

MCE Membrane Filter, 5.0 μm Pore Size

greener alternative

MF-Millipore, filter diam. 47 mm, hydrophilic

Synonym(s):

Cellulose nitrate membrane filter, Hydrophilic MCE membrane filter discs, MF-Millipore Membrane Filter, 5 µm pore size, Mixed Cellulose Ester membrane filter discs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
5.0 μm pore size, 84 % porosity
Material:
mixed cellulose esters (MCE) membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
47 mm
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

agency

in accordance with ASTM® D5755

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore, Millipore

sustainability

Greener Alternative Product

parameter

32 L/min-cm2 air flow rate, 580 mL/min-cm2 water flow rate, 75 °C max. temp.

diam.

47 mm

filter diam.

47 mm

thickness

135 μm

gravimetric extractables

6%

refractive index

n/D 1.495

matrix

MF-Millipore

pore size

5.0 μm pore size, 84 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥0.42 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

General description

  • MF-Millipore® membrane filters are Mixed Cellulose Esters (MCE) of cellulose acetate and cellulose nitrate.
  • Ideal for use in biological and environmental monitoring and various analytical/research applications.
  • Filters are compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods.
  • Available in various formats and pore sizes to suit your application needs.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.
Need a custom membrane filter? Click here to request a custom membrane filter or unique and large package sizes that may not be available via our website.

Application

  • QC of fluid holding tanks
  • Fluid monitoring
  • Particle collection
  • Particle analysis

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


pictograms

Flame

signalword

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Classifications

ED ENV 1 - Flam. Sol. 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library