Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.20649

Sigma-Aldrich

Hippuric acid

for synthesis

Synonym(s):

Hippuric acid, Benzamidoacetic acid, Benzoylglycine, Benzoylaminoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO3
CAS Number:
Molecular Weight:
179.17
MDL number:
UNSPSC Code:
12352209
EC Index Number:
207-806-3
NACRES:
NA.22

Quality Level

Assay

≥97.5% (acidimetric)

form

solid

pH

2.5-3.5 (20 °C, 10 g/L in H2O, slurry)

mp

188-190 °C

solubility

3.26 g/L

bulk density

630 kg/m3

storage temp.

2-30°C

InChI

1S/C9H9NO3/c11-8(12)6-10-9(13)7-4-2-1-3-5-7/h1-5H,6H2,(H,10,13)(H,11,12)

InChI key

QIAFMBKCNZACKA-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 97.5 %
Melting range (lower value): ≥ 186 °C
Melting range (upper value): ≤ 190 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service