Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20276

Sigma-Aldrich

2-Chlorobenzaldehyde

for synthesis

Synonym(s):

2-Chlorobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClO
CAS Number:
Molecular Weight:
140.57
MDL number:
UNSPSC Code:
12352114
EC Index Number:
201-956-3
NACRES:
NA.22

vapor pressure

1.7 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

397 °C

potency

2160 mg/kg LD50, oral (Rat)

pH

2.9 ( in H2O, saturated aqueous solution)

mp

10 °C

transition temp

flash point 87 °C

solubility

1.8 g/L

density

1.25 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1c(cccc1)C=O

InChI

1S/C7H5ClO/c8-7-4-2-1-3-6(7)5-9/h1-5H

InChI key

FPYUJUBAXZAQNL-UHFFFAOYSA-N

Application

  • Synthesis, structural and spectroscopic properties of Co (II), Ni (II) and Cu (II) complexes with 2-((2-chlorobenzylidene) amino) acetohydrazide hydrate and their Antimicrobial and antioxidant activities: Focuses on the synthesis of metal complexes using 2-chlorobenzaldehyde for enhanced antimicrobial and antioxidant properties (FM Al-Azab, Y Mosa′d Jamil, AA Al-Gaadbi, 2023). Link to article
  • Synthesis and Aminomethylation of 2-Amino-4-(2-chlorophenyl)-6-(dicyanomethyl)-1,4-dihydropyridine-3,5-dicarbonitrile N-Methylmorpholinium Salt: Discusses the synthesis involving 2-chlorobenzaldehyde in the production of novel organic compounds (AO Kurskova, VV Dotsenko, KA Frolov, 2022). Link to article
  • Metal Complexes of Schiff Bases Prepared from Quinoline-3-Carbohydrazide with 2-Nitrobenzaldehyde, 2-Chlorobenzaldehyde and 2, 4-Dinitrobenzaldehyde: Studies the synthesis and spectral analysis of Schiff base metal complexes incorporating 2-chlorobenzaldehyde (M Sunjuk, L Al-Najjar, M Shtaiwi, BI El-Eswed, 2023). Link to article
  • Application of 2-chlorobenzaldehyde thiocarbohydrazone as a chromogenic ligand for cadmium (II) detection and removal from water and food: Explores the use of a derivative of 2-chlorobenzaldehyde as a sensitive reagent for detecting and removing cadmium from various samples (AV Sadlapurkar, UB Barache, AB Shaikh, 2024). Link to article

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.248 - 1.250
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service