Skip to Content
MilliporeSigma
All Photos(1)

Documents

820026

Sigma-Aldrich

Probumin® Fatty Acid Free

≥98%, Serology: suitable, enzyme immunoassay: suitable

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41141801
NACRES:
NA.25

product name

Probumin® Fatty Acid Free, pkg of 5 kg

grade

special quality (Specialty Grade)

Quality Level

Assay

≥98% (purity)

form

lyophilized powder

shelf life

48 mo.

mol wt

avg mol wt 66 kDa

purified by

heat shock fractionation

packaging

pkg of 5 kg

manufacturer/tradename

Probumin®

origin

USA origin

free fatty acid content content

0.0-0.2 (mg/g)

technique(s)

enzyme immunoassay: suitable

dilution

(Serology)

loss

≤5%

pH range

6.8-7.2

anion traces

chloride (Cl-): 0.0-6.0 mg/g

cation traces

Na: 0.0-10.0 mg/g

foreign activity

Bluetongue, none detected
Vesicular Stomatitis Virus, none detected

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service