Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.16142

Sigma-Aldrich

Dodecyltrimethylammonium chloride

for synthesis

Synonym(s):

Dodecyltrimethylammonium chloride, Lauryl trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H34ClN
CAS Number:
Molecular Weight:
263.89
MDL number:
UNSPSC Code:
12352107
EC Index Number:
203-927-0
NACRES:
NA.22

Quality Level

form

solid

potency

200-1000 mg/kg LD50, oral (Rat)

mp

246 °C (decomposition)

density

1 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Cl-].[N+](CCCCCCCCCCCC)(C)(C)C

InChI

1S/C15H34N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1

InChI key

DDXLVDQZPFLQMZ-UHFFFAOYSA-M

Linkage

Replaces: DX2477

Analysis Note

Assay (argentometric): ≥ 97.0 %
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service