Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.14688

Sigma-Aldrich

Benzyl chloroformate

(stabilised) for synthesis

Synonym(s):

Benzyl chloroformate, Benzyl chlorocarbonate, Carbonochloride acid benzylester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO2
CAS Number:
Molecular Weight:
170.59
MDL number:
UNSPSC Code:
12352108
EC Index Number:
207-925-0
NACRES:
NA.22

vapor pressure

1 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (acidimetric)

form

liquid

autoignition temp.

590 °C

potency

3000 mg/kg LD50, oral (Rat)

bp

103 °C/25 hPa

mp

-18 °C

transition temp

flash point 92 °C

density

1.21 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C8H7ClO2/c9-8(10)11-6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

HSDAJNMJOMSNEV-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 96.0 %
Density (d 20 °C/ 4 °C): 1.205 - 1.215
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

197.6 °F

Flash Point(C)

92 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service