Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.02353

Sigma-Aldrich

Phenyl chloroformate

for synthesis

Synonym(s):

Phenyl chloroformate, Chloroformic acid phenyl ester, Phenyl chlorocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClO2
CAS Number:
Molecular Weight:
156.57
MDL number:
UNSPSC Code:
12352108
EC Index Number:
217-547-8
NACRES:
NA.22
grade:
for synthesis
application(s):
peptide synthesis
bp:
188-189 °C/1013 hPa
vapor pressure:
0.9 hPa ( 20 °C)

grade

for synthesis

Quality Level

vapor pressure

0.9 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

540 °C

potency

1730 mg/kg LD50, oral (Rat)
4880 mg/kg LD50, skin (Rabbit)

dilution

(for synthesis)

bp

188-189 °C/1013 hPa

mp

-28 °C

transition temp

flash point 69 °C

density

1.24 g/cm3 at 20 °C

application(s)

peptide synthesis

storage temp.

15-25°C

SMILES string

ClC(=O)Oc1ccccc1

InChI

1S/C7H5ClO2/c8-7(9)10-6-4-2-1-3-5-6/h1-5H

InChI key

AHWALFGBDFAJAI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.240 - 1.245
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service