Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14587

Sigma-Aldrich

Triethylene glycol monomethyl ether

for synthesis

Synonym(s):

Triethylene glycol monomethyl ether, Methyltriglycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16O4
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-962-1
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

210 °C

potency

11850 mg/kg LD50, oral (Rat)
7448 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C in H2O)

bp

245-255 °C/1013 hPa

transition temp

flash point 133 °C

density

1.04 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H16O4/c1-9-4-5-11-7-6-10-3-2-8/h8H,2-7H2,1H3

InChI key

JLGLQAWTXXGVEM-UHFFFAOYSA-N

Application

<ul>
<li><strong>Environmental Monitoring</strong>: Nitrite tests are widely used in environmental monitoring to measure nitrite levels in water and wastewater. These tests help assess the impact of agricultural runoff, industrial discharges, and wastewater treatment processes on water quality. An advanced spectrophotometric method using mesoporous silica nanospheres has been developed for the sensitive and rapid determination of nitrite in wastewater, demonstrating its importance in environmental protection (Abou-Melha, ChemistrySelect, 2020).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.047 - 1.049
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service