Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

484245

Sigma-Aldrich

Tri(propylene glycol) methyl ether, mixture of isomers

≥97.5%

Synonym(s):

m-PEG3-alcohol, Methoxytriglycol, Methyltrioxitol, Tripropylene glycol monomethyl ether, Tripropyleneglycol methyl ether, [2-(2-Methoxymethylethoxy)methylethoxy]propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OC3H6)3OH
CAS Number:
Molecular Weight:
206.28
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥97.5%

refractive index

n20/D 1.43 (lit.)

bp

100 °C/2 mmHg (lit.)

density

0.963 g/mL at 25 °C

SMILES string

O(C(COC(COC)C)C)C(CO)C

InChI

1S/C10H22O4/c1-8(5-11)14-10(3)7-13-9(2)6-12-4/h8-11H,5-7H2,1-4H3

InChI key

GVZNXUAPPLHUOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tri(propylene glycol) methyl ether can be used:
  • As a solvent in resins, coatings, dyes for textiles, leather, and cosmetics.
  • As a fuel additive for minimizing the particulate matter emission.
  • To prepare water-mimicking non-aqueous ionic liquids for enzyme compatibility in the trans-esterification process.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzyme activation by water-mimicking dual-functionalized ionic liquids
Hua Zhao, et al.
Molecular Catalysis, 515, 111882-111882 (2021)
Alfredo A Marchetti et al.
Chemosphere, 52(5), 861-868 (2003-05-22)
The addition of oxygen-bearing compounds to diesel fuel considerably reduces particulate emissions. TGME and DBM have been identified as possible diesel additives based on their physicochemical characteristics and performance in engine tests. Although these compounds will reduce particulate emissions, their
An examination of the physical properties, fate, ecotoxicity and potential environmental risks for a series of propylene glycol ethers
Charles A Staples and John W Davis
Chemosphere, 49(1), 61-73 (2002)
Effects of oxygenates on soot processes in DI diesel engines: experiments and numerical simulations
Mueller, et al.
11th Int. Conf. Adv. Prostagl. Leukot. Res., 964-982 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service