Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08245

Sigma-Aldrich

Triethylene glycol

for synthesis

Synonym(s):

Triethylene glycol, Triglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)3H
CAS Number:
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-953-2
NACRES:
NA.22
Assay:
≥99% (GC)
bp:
285-295 °C/1013 hPa
vapor pressure:
<0.01 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

370 °C

potency

17000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

expl. lim.

0.9-9.2 % (v/v)

pH

6.5-7.5 (20 °C, 100 g/L in H2O)

bp

285-295 °C/1013 hPa

mp

-7 °C

transition temp

flash point 165 °C

density

1.123 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOCCO)CCO

InChI

1S/C6H14O4/c7-1-3-9-5-6-10-4-2-8/h7-8H,1-6H2

InChI key

ZIBGPFATKBEMQZ-UHFFFAOYSA-N

Application


  • Elucidating the Role of Reduction Kinetics in the Phase-Controlled Growth on Preformed Nanocrystal Seeds: A Case Study of Ru: This study explores the phase-controlled growth of ruthenium nanocrystals using triethylene glycol as a reducing agent, providing insights into the kinetics of nanomaterial synthesis for catalytic applications (Nguyen et al., 2024).

  • Deep Eutectic Solvents as Agents for Improving the Solubility of Edaravone: Experimental and Theoretical Considerations: This research investigates the use of deep eutectic solvents, including those formulated with triethylene glycol, to enhance the solubility of edaravone. The findings demonstrate significant improvements in drug solubility and bioavailability (Jeliński et al., 2024).

  • Probing the evaporation dynamics of semi-volatile organic compounds to reveal the thermodynamics of liquid-liquid phase separated aerosol: This study uses triethylene glycol in experiments to understand the evaporation dynamics and thermodynamics of semi-volatile organic compounds in liquid-liquid phase-separated aerosols. The research provides valuable data for atmospheric chemistry and environmental science (Choczynski et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.123 - 1.124
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

330.8 °F - closed cup

Flash Point(C)

166 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service