Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.06021

Sigma-Aldrich

Methanesulfonyl chloride

for synthesis

Synonym(s):

Methanesulfonyl chloride, Methanesulfonyl chloride, Mesyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
114.55
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-706-1
NACRES:
NA.22

vapor pressure

2.79 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

435 °C

potency

205-317 mg/kg LD50, oral (Rat)
200-2000 mg/kg LD50, skin (Rabbit)

mp

-32 °C

transition temp

flash point 102 °C

density

1.478 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/CH3ClO2S/c1-5(2,3)4/h1H3

InChI key

QARBMVPHQWIHKH-UHFFFAOYSA-N

General description

for synthesis

Application

Methanesulfonyl chloride for synthesis. CAS 124-63-0, EC Number 204-706-1, chemical formula CH₃SO₂Cl.

Analysis Note

Assay (GC, area%)≥ 99.0 % (a/a)Density (d 20 °C/ 4 °C)1.473 - 1.476Identity (IR)passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service