Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07994

Sigma-Aldrich

3-Sulfolene

for synthesis

Synonym(s):

3-Sulfolene, 2,5-Dihydrothiophene 1,1-dioxide, Butadiene sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2S
CAS Number:
Molecular Weight:
118.15
MDL number:
UNSPSC Code:
12352119
EC Index Number:
201-059-7
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

crystals

potency

2547 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

mp

62-65 °C

transition temp

flash point 112 °C

solubility

130 g/L

density

1.31 g/cm3 at 15.6 °C

bulk density

840 kg/m3

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)CC=CC1

InChI

1S/C4H6O2S/c5-7(6)3-1-2-4-7/h1-2H,3-4H2

InChI key

MBDNRNMVTZADMQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 63 °C
Melting range (upper value): ≤ 66 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service